Learn More
Keystone Cleanroom Connection Shoe Cover
SDP
Catalog No. 18366658 Shop All New Pig Corporation Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-366-658 Large
18-366-659 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-366-658 Supplier Keystone Supplier No. WPL980L
Add to Cart
Add to Cart
Water-repellent CPE plastic shoe covers for cleanrooms to protect your shoes from contamination, splashes, and spills.
  • Overshoe for cleanrooms as well as chemotherapy-related tasks
  • CPE plastic shoe covers are water-repellent for when liquid splash protection is important
  • ISO6 Class 1000+ compatible
  • Snug-fitting to prevent trip hazard; can be used as under-layer when double covering shoes
  • Durable to rely on while working
  • Lint-free, nothing to shed, keeping your cleanroom free of contaminants
  • Latex-free to avoid possible allergic reactions
  • 50 pair per package
Specifications
Size Large
Material Polyethylene
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Plain
Resistance Water, Splash
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 1000 (ISO 6)