Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Overshoe for cleanrooms as well as chemotherapy-related tasks
- CPE plastic shoe covers are water-repellent for when liquid splash protection is important
- ISO6 Class 1000+ compatible
- Snug-fitting to prevent trip hazard; can be used as under-layer when double covering shoes
- Durable to rely on while working
- Lint-free, nothing to shed, keeping your cleanroom free of contaminants
- Latex-free to avoid possible allergic reactions
- 50 pair per package
Specifications
Specifications
| Size | X-Large |
| Material | Polyethylene |
| Latex Free | Yes |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Plain |
| Resistance | Water, Splash |
| Product Type | Controlled Environments Shoe Covers |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
