Learn More
Fisherbrand™ Cleanroom Dry Erase White-Board Marker, Chisel Tip
Catalog No. 18003120 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Blue
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-003-120 Red
18-003-118 Black
18-003-119 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-003-120
Only null left
Add to Cart
Add to Cart
Cleanroom dry erase white-board marker with chisel tip.
  • Cleanroom dry erase marker, chisel tip.
  • Our Cleanroom Dry Erase Whiteboard markers are Class 10 certified and are made with an alcohol base for low odor.
  • These dry erase markers work on whiteboards and most non-porous surfaces.
  • They come in packs of 12 and are double bagged.
  • The ink can be erased without solution using a dry wipe.
Specifications
Material Plastic
Color Red
Product Type Cleanroom Dry Erase Marker