Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURUS™ Cleanroom Shoes

Catalog No. 19125958 Shop All Purus International Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-125-958 X-Large
19-125-954 X-Small
19-125-955 Small
19-125-956 Medium
19-125-957 Large
19-125-959 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-125-958 Supplier PURUS™ Supplier No. CRSHWXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

PURUS™ Cleanroom Shoes provide comfort and durability when working in cleanroom environments.

  • Smooth surface for day-long comfort
  • Antistatic
  • Autoclavable
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanroom
  • Non-slip sole
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Anatomic Heel Basin
  • Toe Grabber

Specifications

Certifications/Compliance ASTM F2913
Color White
For Use With (Application) Pharmaceutical, Biotech, Semiconductor, Cleanrooms, Hospitals
Material Thermoplastic Elastomer
Size X-Large
Toe Style Plain Toe
Packaging Quantity 6/Cs.
Product Type Cleanroom Shoes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.