Learn More
Poltex Cleanroom Sign Sleeve

Description
- PETG sign sleeve.
- Suitable for clean room applications.
- Front face lifts off for easy, thorough cleaning.
- Choose from screw mount, magnet mount, or VHB Tape mount.
Sign sleeve, cleanroom, clean room, Poltex
Specifications
Specifications
| Material | Polycarbonate |
| Size (English) | 8.5 x 11 in. |
| Mounting Type | Self-adhesive |
| For Use With (Application) | Clean Room Applications |
| Shape | Rectangular |
| Languages | Universal |
| Product Type | Sign Sleeve |
| Color | Clear |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.