Learn More
S-Curve™ Cleanroom Sock Dispenser with 2 Compartments
Catalog No. 19082064 Shop All S-Curve Technologies Products
Change view
Click to view available options
No. of Compartments:
2
Catalog No. No. of Compartments
19-082-064 2
1 options
Catalog No. 19-082-064 Supplier S-Curve™ Supplier No. FSD200PETG
Add to Cart
Add to Cart
Large Capacity Cleanroom Sock Dispenser with 2 compartments and front access holes. Clear PETG material is chemical and impact resistant.
A Two Compartment dispenser for Cleanroom Socks.
  • 2 compartment dispenser with front access holes
  • 1/4 in. clear PETG material, SAFE with IPA
  • mounts to wall or free-standing
  • flame treated edges for a smooth finish
  • 1 per case
  • Size: 15.5 x 24 x 11.5 in.
Specifications
Product Type Dispenser
Color Clear
Dimensions (L x W x H) 15.5 x 24 x 11.5 in.
Holds PPE
Material PETG
Mounting Freestanding or Wall Mount
No. of Compartments 2
Product Style Hinged Lid with Round Front Opening
Includes 2 compartments
Sterility Non-sterile