Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purus™ Pure Comfort Cleanroom Socks
GREENER_CHOICE

Catalog No. 19125961 Shop All Purus International Inc. Products
Change view
Click to view available options
Disposable:
Reusable
Single-use
Size:
Large
Medium
One Size
Small
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Disposable Size
19-125-961 Reusable Small
17-456-039 Single-use One Size
19-125-962 Reusable Medium
19-125-963 Reusable Large
19-125-964 Reusable X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-125-961 Supplier Purus™ Supplier No. CRSLS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Purus™ Cleanroom Socks are designed for cleanroom use and comfort.

  • Available in both disposable and launderable styles
  • Made from 100% polyester
  • Cleanroom compatible
  • Comfortable for extended use
  • No particle generation

Specifications

Color White
For Use With (Application) Pharmaceutical, Biotechnology, Cleanrooms, Hospital, Semiconductors
Material Polyester
Size Small
Disposable Reusable
Sterility Non-sterile
Packaging Quantity 1 Pair/Bag, 200 Pair/Cs
Product Type Launderable Cleanroom Socks
Cleanroom Class Class 10-1000 (ISO 4-6)
Packaging Type 200 Pair/Cs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.