Learn More
Fisherbrand™ Cleanroom Sticky Note
Catalog No. 18003103 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Canary
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-003-103 Canary
18-003-104 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-003-103
Only null left
Add to Cart
Add to Cart
Cleanroom sticky note. Does not leave residue and is safe in controlled environments. 3 x 3 in.
  • Class 10 (3x3 in.) 28# custom cleanroom clean notes have 100 latex sheets and are designed for use in critical environments where low particulation and ESD's are required.
  • The latex sheet is smudge-resistant and the adhesive used for binding the sheets does not emit ESD's or carry particulates when a sheet is removed.
Specifications
Material Paper
Color Canary
Product Type Cleanroom Sticky Notes