Learn More
CleanSpace™ CleanSpace2™ Particulate Filter HEPA Hi Capacity
Shop All Cleanspace Technology ProductsDescription
- Compatible with ALL CleanSpace™PAPR Power Units
- CleanSpace™High Efficiency (HEPA) Particulate Filters High Capacity must be used in conjunction with CleanSpace™Filter Adaptor (PAF-0038)
- Used with the revolutionary CleanSpace™- A light weight PAPR with no hoses/belts
- Suitable for protection against airborne particulate (dust, mists and fumes).
- Materials: Fibreglass particulate media and plastic casing, silicone seal
- Easily fitted and removed from the Power Unit
Specifications
Specifications
| Product Type | Filter |
| For Use With (Equipment) | PAF-1034 |