Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Cleanup Kit Variety Pack
SDP

Catalog No. 18366572 Shop All New Pig Corporation Products
Change view
Click to view available options
Absorbancy:
55.3 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Absorbancy
18-366-572 55.3 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-572 Supplier PIG™ Supplier No. KIT5018
Add to Cart
Add to Cart

Variety pack of single-use spill kits.

  • Compact design lets you store a spill kit virtually anywhere you need one for fast response
  • Easy-open bag protects kit contents from dirt and moisture
  • Absorbs oils, coolants, solvents, water, universal, acids, bases, unknowns, bodily fluids, urine
  • Includes: 6 - 10 x 13 in. lightweight universal absorbent mat pads, 3 - 10 x 13 in. lightweight HazMat chemical absorbent mat pads, 2 - 3-5 oz. pouches loose absorbent, 1 - 3.5 oz. pouch acid encapsulating/neutralizing absorbent, 1 - 3.5 oz. pouch base encapsulating/neutralizing absorbent, 3 - pairs of XL nitrile gloves, 1 - 18 x 30 in. polyethylene disposal bag, 1 - 33 x 39 in. biohazard waste bag, 1 - 8 x 12 in. premoistened hand wipe, 1 - 7.5 x .25 in. nylon zip tie, 3 - disposable scoops with scraper, 3 - resealable poly bags
  • Spill Kit dimensions are approximate

Specifications

Absorbancy 55.3 oz.
Material Polypropylene
Type Spill Kit
Weight (English) 3 lb.
Weight (Metric) 1.4 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.