Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CleanWear™ Tyvek™ Disposable Coveralls with Elastic Ankles and Wrists

Catalog No. 18892403 Shop All Cleanwear USA Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-892-403 Large
18-892-401 Small
18-892-402 Medium
18-892-404 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-892-403 Supplier CleanWear™ Supplier No. 1154LG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tough coverall protects skin from wet and dry particulate matter

Provides protection from a variety of harmful materials including paint, chemicals, construction products, oil, grease, fiberglass, mold, etc.

  • Microscopic particles as small as 0.5μm are unable to pass through Tyvek even if the fabric has been abraded
  • Not easily scratched or worn away
  • Made with one material in one layer, unlike laminated fabrics that leave you exposed if the outer coating is scratched off
  • Let air and moisture vapor pass through, reducing the chances of heat stress
  • Six times more breathable than microporous materials
  • Disposable
  • Elastic ankles and wrists protect arms and legs from exposure to harmful or irritating matter

Specifications

Product Type Cleanroom Coveralls
Size Large
Material Tyvek Spunbond Olefin
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Description Elastic wrist and ankle, collar
For Use With (Application) Barrier protection in controlled environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.