Learn More
Eisco™ Clear Acrylic Rod
SCI_ED
Catalog No. S27756 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27756 1 Ea.
1 options
Catalog No. S27756 Supplier Eisco™ Supplier No. PH1001X1
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
When light is transmitted through this clear acrylic rod the effect is similar to that of fiber optic lighting
  • Transmits light similar to fiber optics
  • Made of clear, hard acrylic
  • Width: 1 in.
Specifications
Product Type Rod
Description 1 x 1 in.
Material Acrylic
For Use With (Application) Light Transmission
Quantity 1 Ea.
Height (English) 1 in.