Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Clear Screw Caps

Catalog No. MP115068005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 Pk.
500 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP115068005 1000 Pk.
MP115068002 500 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP115068005 Supplier MP Biomedicals™ Supplier No. 115068005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use with empty FastPrep™ tubes. Clear Caps are screw scaps with an O-ring for use with 2 mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix tubes for prepare samples for homogenization

  • Clear Caps fit empty FastPrep™ tubes and are used in FastPrep systems.
  • Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap.
  • Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Volume (Metric) 2 mL
Storage Requirements Store at Room Temperature (15°C to 30°C)
Quantity 1000 Pk.
Product Type Screw Cap
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.