Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Molded Glass Type I Borosilicate Serum Vials

Catalog No. 01672066
Change view
Click to view available options
Volume (Metric):
20 mL
50 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-672-066 50 mL
01-672-069 100 mL
01-672-070 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-672-066 Supplier Qorpak™ Supplier No. 327208

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

The utilization of type I molded glass enhances mechanical resilience, thereby minimizing the potential for particle contamination during crucial fill/finish and lyophilization stages. Vials are depyrogenated and sterilized. Crimp cap not included.

  • 100% inspected in clean room environment
  • Vials are washed with water for injection and dried in an ISO8 environment
  • Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
  • Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
  • Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
  • Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal

Specifications

Color Clear
Diameter (Metric) Outer 42.5 mm
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Disposable Single-use
Volume (Metric) 50 mL
For Use With (Application) Lyophilization
Graduated No
No. per Case 168/Cs.
No. per Pack 28/Pk.
Packaging Type Tray Packed Neck Up
Size (Metric) Opening 20 mm
Dnase Free No
Closure Type Crimp Cap
Quantity 168 Vials
Marking Spot No
Fit for Purpose Sterile
Vial Type Ready to Use/Sterile
Autoclavable Autoclavable
Height (Metric) 73 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.