Learn More
Analytical Sales & Services Clear Polypropylene Reservoir, 25pk
Supplier: Analytical Sales & Services 21012
Clear Polypropylene Reservoir 12 Channel Deep Well Partitioned 21mL/Channel 252mL Plate Volume 96 Well Format
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.