Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Clear Vinyl Bib Apron
SCI_ED

Catalog No. p-9451000 Shop All Eisco Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S24922 Medium
S24921 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S24922 Supplier Eisco™ Supplier No. VNAPCH
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clear vinyl bib apron protects clothes during laboratory exercises. Ideal for use in labs, kitchens, or shops.

  • Basic transparent vinyl apron
  • Perfect for biology, chemistry, and general science laboratories
  • Protects clothes from liquid spills and splashes
  • Acid resistant

Specifications

Type Bib Apron
Size Medium
Material Vinyl
Color Clear
Disposable Reusable
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
Thickness 6.7 mil (0.17 mm)
For Use With (Application) Labs, kitchens or shops
Seam Type Seamless
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.