Learn More
StatLab CLEARIFY CLEARING AGENT
SDP
Supplier:  StatLab CACLEGAL
Encompass_Preferred
Clearify is an amazing xylene alternative that is biodegradable, low-hazard, and non-flammable (Flash Point 150F). This product is non-carcinogenic, non-dermatologic, and non-corrosive.
Catalog No. NC9837230
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.