Learn More
Invitrogen™ CLEC12A Monoclonal Antibody (687317)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | CLEC12A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 687317 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CLEC12A |
| Gene Accession No. | Q5QGZ9 |
| Gene Alias | CD371; Clec12a; CLL1; CLL-1; C-type lectin domain family 12 member A; C-type lectin domain family 12, member A; C-type lectin protein CLL-1; C-type lectin-like molecule 1; C-type lectin-like molecule-1; D230024O04; DCAL2; DCAL-2; Dendritic cell-associated lectin 2; KLRL1; Micl; Myeloid inhibitory C-type lectin-like receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.