Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142472
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142472 Supplier Thermo Scientific Supplier No. PA5142472
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

CLEC2D Polyclonal Antibody

CLEC2D Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC2D
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CLEC2D
Gene Accession No. Q9UHP7
Gene Alias CLAX; Clec2d; Clrb; Clr-b; C-type lectin domain family 2 member D; C-type lectin domain family 2, member d; C-type lectin related f; C-type lectin superfamily 2, member D; C-type lectin-domain family 2 member D; C-type lectin-related protein B; lectin-like NK cell receptor; Lectin-like transcript 1; lectin-like transmembrane protein; LLT1; LLT-1; MGC123478; OCIL; Osteoclast inhibitory lectin
Gene Symbols CLEC2D
Host Species Goat
Immunogen Peptide with sequence C-LLRYKGPSDH.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29121
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.