Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4E Monoclonal Antibody (OTI3F4), TrueMAB™, OriGene
SDP

Catalog No. p-200011725
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9877 30 μL
50-167-5698 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9877 Supplier OriGene Supplier No. TA505160S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CLEC4E Monoclonal antibody specifically detects CLEC4E in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen CLEC4E
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PLPBP
Gene Accession No. O94903
Gene Alias C86253; CLEC4E; CLECSF9; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 9; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 9; C-type lectin domain family 4 member E; C-type lectin domain family 4, member E; C-type lectin superfamily member 9; C-type lectin, superfamily member 9; immunoreceptor; macrophage-inducible C-type lectin; Mincle; UNQ218/PRO244
Host Species Mouse
Immunogen Full length human recombinant protein of human PROSC produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11212
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.