Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLEC4F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547396 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547396 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547396 Supplier Invitrogen™ Supplier No. PA547396
Add to Cart
Edge
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CLEC4D, rhCLEC4E, and rhCLEC4F is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Receptor with an affinity for galactose and fucose. Could be involved in endocytosis. Target protein's carbohydrate binding receptor with an affinity for fucose and galactose.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLEC4F
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Clec4f
Gene Accession No. P70194
Gene Alias Clec4f; Clecsf13; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 13; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13; C-type lectin 13; C-type lectin domain family 4 member F; C-type lectin domain family 4, member f; C-type lectin superfamily member 13; D18063; KCLR; KCR; KUCR_MOUSE; Kupffer cell c-type lectin receptor; kupffer cell receptor
Gene Symbols Clec4f
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CLEC4F/CLECSF13 Ala65-Gly548.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51811
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.