Learn More
Apexbio Technology LLC Fexofenadine HCl 153439-40-8 10mM (in 1mL DMSO)
Supplier: Apexbio Technology LLC B15735.1
Fexofenadine HCl (CAS 153439-40-8) is a non-sedating antihistamine that selectively antagonizes the histamine H1 receptor It inhibits H1 receptor-mediated signaling with an IC50 of 246 nM indicating potent receptor binding in vitro Fexofenadine is employed in biomedical research to investigate histaminergic pathways allergic responses and receptor pharmacology facilitating the study of H1 receptor antagonism in cellular and molecular models
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.