Learn More
Apexbio Technology LLC Clemastine Fumarate 14976-57-9 500mg
SDP
Supplier:  Apexbio Technology LLC B1558500
Encompass_Preferred
Clemastine Fumarate (CAS 14976-57-9) is a small molecule that functions as a selective antagonist of the histamine H1 receptor It demonstrates high affinity for the receptor with an inhibitory concentration (IC50) of 3 nM By blocking histamine H1 receptor-mediated signaling clemastine fumarate is utilized in research to study histaminergic pathways and allergic responses It is also employed to investigate the pharmacological modulation of H1 receptor activity in various cellular and in vivo models
Catalog No. 50-000-90608
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.