Learn More
Medchemexpress LLC HY-B0298A 500mg , Clemastine (fumarate) CAS:14976-57-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0298A500MG
Encompass_Preferred
HY-B0298A 500mg Clemastine (fumarate) CAS: 14976-57-9 Clemastine (HS-592) fumarate is a selective histamine H1 receptor antagonist. Clemastine fumarate is an antihistamine mainly used for relieving symptoms of allergic reactions primarily by competing with histamine to bind H1 receptors. Anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1125
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.