Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Clemizole hydrochloride
SDP

Catalog No. 537150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-715-0 50 mg
53-711-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-715-0 Supplier Tocris Bioscience™ Supplier No. 5371/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

TRPC5 blocker; also H1 antagonist

TRPC5 channel blocker (IC50 values are 1.1, 6.4, 9.1, 11.3 and 26.5 μM for TRPC5, TRPC4, TRPC3, TRPC6 and TRPC7 respectively). Exhibits minimal effect on TRPM3, TRPM8 and TRPV1-4 channels. Also histamine H1 receptor antagonist and anticonvulsant in zebrafish Dravet syndrome model.

Specifications

Purity >99%
Quantity 50 mg
Product Type Clemizole hydrochloride
Molecular Weight (g/mol) 362.3
Inhibitors Clemizole hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.