Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clever Culture Systems APAS™ Independence Instrument

Catalog No. R16001 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
100-240 VAC, 50∼60Hz, 6 Amps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
R16001 100-240 VAC, 50∼60Hz, 6 Amps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R16001 Supplier Thermo Scientific Supplier No. R16001

This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Maximize efficiency with the first automated culture plate reader APAS Independence brings clever automation to laboratories, freeing up valuable microbiologist time to focus on other tasks.

Maximize efficiency with the first automated culture plate reader. Free up valuable tech time for more meaningful activities with the APAS Independence instrument (Automated Plate Assessment System).

The APAS Independence instrument utilizes the breakthrough Automated Plate Assessment System (APAS) imaging technology and artificial intelligence algorithms to read and categorize agar plates, and remove negatives from the workflow.

Embrace automation in microbiology without the need for large-scale capital resources to streamline with the APAS Independence instrument.

Streamline your workflow

The intuitive nature of the APAS Independence instrument means your workflow remains the same no matter what modules* are used. Training on the Instrument is easy, and module-specific training can be added when required.

Save more than time

Not only does the APAS Independence Instrument save you time by alleviating the bottlenecks in bench reading, it also impacts many aspects of managing an efficient and effective microbiology lab.

  • Budget management: Helps avoid budget overruns with predictable costs year on year.
  • Increased workload: Adding Analysis Modules allow for optimal throughput.
  • Process improvement: Reduce manual handling and contamination risks.
  • Quality and consistency: Reduce natural variation in the manual plate reading process.
  • Staff satisfaction: Utilize highly trained staff on cases that require your expertise.
  • Time management: Improve your teams' efficiency by allowing them to focus on more meaningful activities.
  • Reduced workplace injury: Reduces potential RSI risks and staff downtime costs.

Embrace Automation in Microbiology

The APAS Independence is task-focused, compact, and designed specifically to overcome the bottleneck in manual culture plate reading by automatically triaging plates into three categories: 'significant', 'non-significant or 'negative', and 'for review'.

Unlike large-scale automation solutions, the APAS Independence instrument offers the benefits of standardization, but in a small, affordable, and flexible footprint designed to easily integrate with the majority of laboratories.

TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W x H) 200 x 80 x 160 cm (78.74 x 31.5 x 62.99 in.)
For Use With (Equipment) 100-240 VAC, 50∼60Hz, 6 Amps
Weight (English) 727.5 lb.
Weight (Metric) 330 kg

Availability of product in each country depends on local regulatory marketing authorization status. ©2021 Thermo Fisher Scientific Inc. Clever Culture Systems is the legal manufacturer of the APAS Independence. APAS is a trademark of Clever Culture Systems. All other trademarks are the property of Thermo Fisher Scientific and its subsidiaries. Products are distributed in the U.S. and Europe so uses, applications, and availability of product in each country depend on local regulatory marketing authorization status.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.