Learn More
Health Care Logistics Inc CLICK OPEN AMPULE OPENER
SDP
Supplier:  Health Care Logistics Inc 7371
Encompass
Ampule Opener, The Ampule Opener reduces the chance of finger cuts, Small cutting blade inside scores the ampule stem to help insure a clean break, Length: 2- 5/8 in. (67mm), Diameter: 3/4 in. (19mm)
Catalog No. NC9839770
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.