Learn More
FUJIFILM IRVINE SCIENTIFIC INC CTGrade GMP rh IL-25 100UG
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 50025100UG
Encompass_Preferred
PDGF-BB (Platelet-derived growth factor) comprises a family of homo or heterodimeric growth factors, including PDGF-AA, PDGF-BB, PDGF-CC, and PDGF-DD. Binds to two different transmembrane tyrosine kinase receptors (PDGFR-alpha and PDFGR-beta) Released from degranulating platelets and plays a role in each stage of wound healing Stimulates mitogenicity and chemotaxis of neutrophils, macrophages, fibroblasts, and smooth muscle cells to the wound site Enhances proliferation of fibroblasts and their production of extracellular matrixThe biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
Catalog No. 50-289-2161
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.