Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Clindamycin, Antimicrobial Agent, >99% Purity, Solid, 50 mg

Catalog No. 502706375
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6375 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6375 Supplier Abcam Supplier No. AB14200550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Broad-spectrum antimicrobial agent that acts as a competitive inhibitor of peptide formation in the 50S ribosomal unit.

  • Molecular weight: 425 Da
  • Purity: >99%
  • Form: Solid
  • Activity: Broad spectrum antimicrobial agent; active against Gram-positive and Gram-negative bacteria and selected protozoa; competitive inhibitor of peptide formation in large 50S ribosomal unit; orally active in vivo
  • CAS number: 18323-44-9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule 50S Ribosomal Unit
Content And Storage Store at 4°C.
Form Solid
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Quantity 50 mg
For Use With (Application) Antimicrobial research, Protein synthesis inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.