Learn More
Abcam Clindamycin, Antimicrobial Agent, Solid, 99 Percent Purity, 10 mg
Description
- Broad spectrum antimicrobial agent
- Active against Gram-positive and Gram-negative bacteria and selected protozoa
- Competitive inhibitor of peptide formation in large 50S ribosomal unit
- MW: 425 Da
- Purity: >99%
- Form: Solid
- Orally active in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Ribosome (50S) |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Reactivity | Gram-positive and Gram-negative bacteria, Protozoa |
| Shipping Condition | Ambient |
| Quantity | 10 mg |
| For Use With (Application) | Antimicrobial research, peptide synthesis inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.