Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clindamycin, White Powder, MP Biomedicals™

Catalog No. ICN19116850 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19116850 50 mg
ICN19116810 10 mg
ICN19116880 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19116850 Supplier MP Biomedicals, Inc Supplier No. 0219116850
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21462-39-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-PIPVPTKYSA-N
Synonym (7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
PubChem CID 134129519
IUPAC Name (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin
Physical Form Solid
Color White
Quantity 50 mg
Formula Weight 461.4
Chemical Name or Material Clindamycin
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.