Learn More
Description
- For manual and automated test methodologies
- For use as assayed quality control materials for Glucose, Bilirubin, Ketones, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes, hCG, Microalbumin, Creatinine, and Galactose
- DipScopics™ also contains red blood cells, white blood cells, and crystals for microscopic evaluation of urine sediment
Specifications
Specifications
| Certifications/Compliance | ISO 9000 13485 |
| Content And Storage | 2°C to 8°C |
| For Use With (Equipment) | Dropstrip |
| Type | DropStrip Liquid QC Urinalysis Controls |
| Intended Use | In-vitro diagnostic use, use as assayed quality control materials for Glucose, Bilirubin, Ketones, specific gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes, hCG, Microalbumin, Creatinine, and Galactose |
| Level | 2 |
| Quantity | 2 x 25 mL |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.