Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Clioquinol, Metal Chelator, Ionophore, Solid, 1 g
Description
- MW: 305.5 Da
- Purity: >97%
- Form: Solid
- Function: Metal chelator and ionophore; inhibitor of CLK-1/MCLK1 (aging-associated protein)
- Key biological effects: Iron, copper, and zinc chelation; inhibits Huntington expression from mRNA; proteasome inhibition when complexed with copper; stimulates tumor necrosis factor-α secretion
- Research applications: Neurodegenerative disease models (Alzheimer's, Parkinson's, and Huntington's disease)
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Metals, CLK-1/MCLK1 |
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% clioquinol |
| Form | Solid |
| Formulation | 100% Clioquinol |
| Quantity | 1 g |
| For Use With (Application) | Neurodegenerative disease research, Alzheimer's disease research, Parkinson's disease research, Huntington's disease research, CLK-1/MCLK1 inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?