Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Clioquinol, Metal Chelator and Ionophore, Solid, 100 mg

Catalog No. 502707658
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7658 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7658 Supplier Abcam Supplier No. AB120355100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Metal chelator and ionophore with potential therapeutic applications in neurodegenerative disease models.

  • Molecular Weight: 305.5 Da
  • Purity: >97%
  • Physical Form: Solid
  • Function: Metal chelator and ionophore; inhibitor of CLK-1/MCLK1
  • Biological Activity: Known beneficial effects in animal models of Alzheimer's, Parkinson's and Huntington's disease, including copper, zinc and iron chelation; stimulates tumor necrosis factor-α secretion
  • Storage: Store at ambient temperature

Specifications

Target Molecule Metals, CLK-1/MCLK1, TNF-α
Shipping Condition Ambient
Content And Storage Store at ambient temperature.
Form Solid
Quantity 100 mg
For Use With (Application) Neurodegenerative disease research, metal chelation, ionophore studies, aging-associated protein inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.