Learn More
United Scientific™ Clips For Jointed Glassware, Polypropylene
SDPSCI_ED
Catalog No. S16040 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
Joint Size 14/23
Joint Size 19/26
Joint Size 29/32
Color:
Blue
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
S16040 Joint Size 14/23 Yellow
S15805 Joint Size 29/32 Red
S15847 Joint Size 19/26 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S16040 Supplier United Scientific™ Supplier No. CLJP1423PK10
Add to Cart
Add to Cart
Prevent breakage in the lab by safely clipping jointed glassware together.
  • Plastic conical joint clips are designed to securely holdjoint size glassware together. These are non-corrosive,and will not scratch glassware.
  • Manufactured from polypropylene
  • Outstanding chemical resistance
  • Maximum operating temperature of 135°C
  • Autoclavable
Specifications
Quantity 10/Pk.
Type Joint Clamp
Material Plastic
Size Joint Size 14/23
Color Yellow
Warning: Cancer and reproductive harm