Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ClipTip™ Filtered 384-Format Pipette Tips
GREENER_CHOICE

Catalog No. 04100511
Change view
Click to view available options
Volume (Metric):
12.5 μL
30 μL
125 μL
Tip Style:
Extended Length
Standard 384
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
04-100-511 12.5 μL Extended Length
14-387-987 12.5 μL Standard 384
14-387-990 30 μL Standard 384
14-387-993 125 μL Standard 384
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 04-100-511 Supplier Thermo Scientific™ Supplier No. 94420073
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.

Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovativesnap and seal tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.

Key Features:

  • All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
  • Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
  • Ideal for 384-format microplate applications
  • Filtered tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations

Proven quality

  • Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Versatile Tip Range

  • 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
  • 384-format volumes: 12.5, 30 and 125μL
  • Extended length tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables

Immediate traceability

  • Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert

Note:

Availability may vary by country.

TRUSTED_SUSTAINABILITY

Specifications

Description ClipTip 384 12.5 Ext, Filtered, Sterile, Rack
Volume (Metric) 12.5 μL
Filtered Filtered
Sterility Sterile
Autoclavable Not Autoclavable
Length (English) 1.61 in
Length (Metric) 4.1 cm
Format Rack
Tip Style Extended Length
Packaging 10 x 384 tips/rack
Color Code Pink
For Use With (Equipment) 384-Format Microplate, Thermo Scientific F1-ClipTip
Product Line ClipTip
Fit for Purpose Sterile & PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.