Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
GREENER_CHOICE
Catalog No. 14488011
Change view
Click to view available options
Description:
ClipTip Empty Rack
ClipTip Empty Rack, Large
ClipTip Empty Rack, Medium
ClipTip Empty Rack, Small
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
14-488-011 ClipTip Empty Rack, Medium
14-488-146 ClipTip Empty Rack, Small
04-100-505 ClipTip Empty Rack
14-488-012 ClipTip Empty Rack, Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-488-011 Supplier Thermo Scientific™ Supplier No. 94410519
Add to Cart
Add to Cart
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
  • Small Rack: For use with ClipTip 20 and 50 Pipette Tips
  • Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
  • Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
TRUSTED_SUSTAINABILITY
Specifications
Description ClipTip Empty Rack, Medium
Model ClipTip Empty Rack
For Use With (Equipment) ClipTip 1000 Pipette Tip
Product Line ClipTip