Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLNS1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556906
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556906 Supplier Invitrogen™ Supplier No. PA556906
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFLKSFPPPG PAEGLLRQQP DTEAVLNGKG LGTGTLYIAE SRLSWLDGSG LGFSLEYPTI SLHALSRDRS DCLGEHLYVM VNAKFEEESK EPVADEE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via methylation of histones, rendering chromatin heritably changed in its expressibility. Involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator for PPARG and enhances its adipocyte differentiation-inducing activity; the function seems to involve differential recruitment of acetylated and methylated histone H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLNS1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLNS1A
Gene Accession No. P54105
Gene Alias 2610036D06Rik; 2610100O04Rik; ARPC1A; chloride channel; chloride channel current inducer; chloride channel regulator; chloride channel regulatory protein; chloride channel, nucleotide sensitive 1A; chloride channel, nucleotide-sensitive, 1A; chloride conductance regulatory protein ICln; Chloride ion current inducer protein; chloride nucleotide-sensitive channel 1A; ClCI; Clcni; Clns 1a; CLNS1A; CLNS1B; I(Cln); icln; icln protein; methylosome subunit pICln; pIcIn; pICln; Rcl1; Reticulocyte pICln; reticulocyte protein ICln; SOP2L; SOP2-like protein; swelling dependent chloride channel; swelling-induced chloride conductance regulatory protein; swelling-induced chloride conductance regulatory protein pIcIn
Gene Symbols CLNS1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLNS1A. Recombinant protein control fragment (Product #RP-95782).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1207
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.