Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clobenpropit Dihydrobromide 97.0+%, TCI America™
SDP

Catalog No. C344625MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C344625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C344625MG Supplier TCI America Supplier No. C344625MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145231-35-2
Molecular Formula C14H19Br2ClN4S
Molecular Weight (g/mol) 470.65
MDL Number MFCD00467655
InChI Key JIJQPEZAVLJZBO-UHFFFAOYSA-N
Synonym 3-(1H-Imidazol-4-yl)propyl 4-Chlorobenzylcarbamimidothioate Dihydrobromide
PubChem CID 11213569
ChEBI CHEBI:64165
IUPAC Name N'-[(4-chlorophenyl)methyl]{[3-(1H-imidazol-5-yl)propyl]sulfanyl}methanimidamide dihydrobromide
SMILES Br.Br.NC(SCCCC1=CN=CN1)=NCC1=CC=C(Cl)C=C1

Specifications

Melting Point 205°C
Quantity 25 mg
Physical Form Crystal-Powder at 20°C
Formula Weight 470.65
Percent Purity ≥97.0% (HPLC)
Chemical Name or Material Clobenpropit Dihydrobromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.