Learn More
Description
Clobetasol 17-Propionate USP is a white to cream, crystalline powder. Micronized with a particle size distribution of 99% ≤ 50 μm.
Alternative name(s): clobetasol propionate, Clobetasone propionate
Specifications
Specifications
| Chemical Name or Material | Clobetasol 17-Propionate USP (Micronized) |
| Packaging | Bottle, Amber Glass |
| CAS | 25122-46-7 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 4669738 |
| Quantity | 250 mg |
| Percent Purity | 97 to 102 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
