Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clobetasol Propionate, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18601746 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-601-746 1 g
18-601-747 5 g
18-601-748 25 g
18-601-749 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-746 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C17851GM

C1785, 25122-46-7, C25H32ClFO5

Spectrum™ Chemical Clobetasol Propionate, Micronized, USP is a corticosteroid of the glucocorticoid class used to treat various skin disorders and contact dermatitis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25122-46-7
Molecular Formula C25H32ClFO5

Specifications

Quantity 1 g
Purity 97-102%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.