Learn More
Clofazimine, G-Biosciences

Description
lofazimine is an active phenazine composed of an imino group and aromatic tricyclic heterocycles that is used as an antimycobacterial, anti-inflammatory and anti-cancer compound. It has been shown to be a potent in vivo antimycobacterial agent toward Mycobacterium leprae and in vitro to Mycobacterium avium.
As an anti-inflammatory, clofazimine inhibits reactive oxygen species (ROS) formation within neutrophils by inhibiting myeloperoxidase catalyzed reactions. The formed ROS are responsible for activating tumor necrosis factor alpha and interleukin 8 (IL8), which are inflammatory cytokines.
Specifications
Specifications
| Quantity | 5 g |
| Boiling Point | 210 to 212°C |
| Product Line | Gibco |
| Physical Form | Powder |
| Purity | 98% |
| Molecular Weight (g/mol) | 473.4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.