Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clofibric acid, MP Biomedicals™

Catalog No. ICN15070110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15070110 10 g
ICN15070180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15070110 Supplier MP Biomedicals, Inc Supplier No. 0215070110
Add to Cart
Add to Cart

Chemical Identifiers

CAS 882-09-7
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.645
InChI Key TXCGAZHTZHNUAI-UHFFFAOYSA-N
Synonym clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
PubChem CID 2797
ChEBI CHEBI:34648
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl

Specifications

Quantity 10 g
Formula Weight 214.6
Physical Form Solid
Chemical Name or Material Clofibric acid
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.