Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Clopamide, Na+/2Cl- Cotransporter Inhibitor, Diuretic, Solid, 500 mg

Catalog No. 502706529
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6529 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6529 Supplier Abcam Supplier No. AB141522500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Selective Na+/2Cl- cotransporter inhibitor with diuretic activity in vivo.

  • Activity: Selective Na+/2Cl- cotransporter inhibitor and diuretic
  • Purity: >98%
  • Molecular Weight: 345.8 Da
  • Form: Solid
  • Administration: Orally active
  • In vivo activity: Diuretic activity

Specifications

Content And Storage Store at ambient temperature.
Target Molecule Na+/2Cl- Cotransporter
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Formulation 100% Clopamide
Quantity 500 mg
For Use With (Application) Na+/2Cl- cotransporter inhibition, Diuretic activity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.