Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clopidol Monoclonal Antibody (3H3)
GREENER_CHOICE

Catalog No. PIMA549720
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549720 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549720 Supplier Invitrogen™ Supplier No. MA549720
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Clopidol is a veterinary drug that belongs to the chemical class of nitrobenzaldehydes. It is commonly used as an anticoccidial agent in poultry farming, which helps to control and prevent the spread of the parasitic disease coccidiosis. Clopidol works by inhibiting the growth and replication of the coccidia parasites in the gut of the birds. It is usually administered orally in the feed or drinking water of the poultry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clopidol
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3H3
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of CLOP hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.