Learn More
ATCC Clostridioides difficile; Strain 4118
Supplier:  ATCC BAA1870
Encompass
Clostridioides difficile strain 4118 is a whole-genome sequenced, anaerobic bacterium. This strain is toxigenic (toxinotype IIIb) and is confirmed by PCR to have the cdtB, tcdA, and tcdB genes. This product has applications in enteric disease and infectious disease research.
Catalog No. 50-238-4776
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.