Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium botulinum Toxin A Recombinant Mouse Monoclonal Antibody (Aa1)

Catalog No. MA554296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA554296 Supplier Invitrogen™ Supplier No. MA554296
Add to Cart
Edge
Add to Cart

Mouse Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

Clostridium botulinum is a gram-positive, anaerobic motile bacteria normally found in soil. C. botulinum is a potential threat to both humans and animals through contaminated food because of the potent neurotoxin they produce. C. botulinum bacteria are categorized within 7 strains A-G based on the characteristics of their toxin. A,B, E and F strains are known to cause food poisoning in humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium botulinum Toxin A
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone Aa1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias botulinum; botulinum toxin; C. botulinum
Host Species Mouse
Immunogen Recombinant Clostridium botulinum botA/BOTOX.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a mFc VHH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.