Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium difficile Glutamate Dehydrogenase (GDH) Chicken anti-Bacteria, Polyclonal, Gallus
SDP

Catalog No. 502100604
Encompass
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-0604 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-0604 Supplier Vector Laboratories Supplier No. ACDGDH
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chicken Polyclonal Antibody

Clostridium difficile Glutamate Dehydrogenase (GDH) Polyclonal antibody specifically detects Clostridium difficile Glutamate Dehydrogenase (GDH) in Bacteria samples. It is validated for ELISA, Western Blot

Specifications

Antigen Clostridium difficile Glutamate Dehydrogenase (GDH)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5-2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.075% sodium azide; pH
Gene Alias Ac2-281; AI118167; brain glutamate dehydrogenase; epididymis tissue sperm binding protein Li 18mP; GDH; GDH 1; GDH1; Gdh-X; GLUD; Glud1; GLUD1 protein; Gludeha; Gludl; glutamate dehydrogenase (NAD(P)+); glutamate dehydrogenase 1; glutamate dehydrogenase 1, mitochondrial; Memory-related gene 2 protein; MRG-2
Host Species Chicken
Immunogen Clostridium difficile Glutamate Dehydrogenase (GDH)
Purification Method Affinity chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.