Learn More
Microbiologics™ Clostridium sporogenes ATCC™ 11437™†
SDP
Catalog No. 23016690 Shop All Microbiologics Products
Change view
Click to view available options
ATCC®:
11437
ATCC 11437
Product Type:
Lyfo-Disk QC Control
Quality Control Organism
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ATCC® Product Type
23-016-690 ATCC 11437 Lyfo-Disk QC Control
23-021182 11437 Quality Control Organism
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-016-690 Supplier Microbiologics™ Supplier No. 0487L
Add to Cart
Add to Cart
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single strain of Clostridium sporogenes in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of microorganism
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 10 pellets
  • ATCC™ 11437™†
Specifications
ATCC® ATCC 11437
Product Type Lyfo-Disk QC Control
Species Clostridium sporogenes
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.