Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clostridium tetani Tetanus Toxin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA173168
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173168 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173168 Supplier Invitrogen™ Supplier No. PA173168
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73168 antibody reacts with Tetanus toxoid. Potential applications for PA1-73168 are numerous including ELISA, Western blot, Immunofluorescence and Immunohistochemical applications. In addition this product may be used in place of neat antiserum in almost any appropriate antibody-based technique. The PA1-73168 antibody was raised against purified tetanus toxoid.

Tetanus toxin is a neurotoxin produced by the vegetative spore of Clostridium tetani. The toxin causes tetanus, a disease that is characterised by prolonged contraction of skeletal muscle. World-wide, approximately 1 million cases of tetanus occurs annually, resulting in approximately 300,000 to 500,000 deaths, mostly in developing countries. The tetanus toxin has a molecular weight of approximately 150kDa and consists of a heavy chain (B-chain, 100kDa) and a light chain (A-chain, 50kDa) that are connected by a disulfide bond. The toxin acts by inhibiting neurotransmitter release. It binds to peripheral neuronal synapses, is internalized and moves by retrograde transport up the axon into the spinal cord where it can move between postsynaptic and presynaptic neurons. It inhibits the release of the neurotransmitters GABA and glycine by synaptobrevin-2. This results in overactivity in the muscle. The binding of the toxin to neurons is irreversible.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clostridium tetani Tetanus Toxin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias C. tetani; Tetanus Toxin
Host Species Goat
Immunogen Purified tetanus toxoid.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.